Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751370.3 | XP_002751416.1 | 6 - 7 | 484 | 85 | |
| XM_009002103.2 | XP_009000351.1 | 7 - 8 | 533 | 85 | ||
| XM_009002104.2 | XP_009000352.1 | 6 - 7 | 643 | 85 | ||
| XM_009002105.2 | XP_009000353.1 | 6 - 7 | 621 | 85 | ||
| XM_009002108.2 | XP_009000356.1 | 7 - 8 | 535 | 85 | ||
| XM_017975151.1 | XP_017830640.1 | 7 - 8 | 529 | 85 | ||
| XM_017975152.1 | XP_017830641.1 | 6 - 7 | 476 | 85 | ||
| XM_017975153.1 | XP_017830642.1 | 7 - 8 | 530 | 85 | ||
More Information
Set Membership: |
|

UBE2D4
Genomic Map