Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748403.3 | XP_002748449.1 | 2 - 3 | 405 | 64 | |
| XM_008997336.1 | XP_008995584.1 | 2 - 3 | 405 | 64 | ||
| XM_008997337.2 | XP_008995585.1 | 2 - 3 | 405 | 64 | ||
| XM_017972633.1 | XP_017828122.1 | 3 - 4 | 444 | 64 | ||
More Information
Set Membership: |
|

MMD
Genomic Map