Assay Details
| Gene Symbol: ASAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979214.1 | XP_008977462.1 | 4 - 5 | 542 | 71 | |
| XM_017963727.1 | XP_017819216.1 | 4 - 5 | 584 | 71 | ||
More Information
Set Membership: |
|

ASAH1
Genomic Map