Assay Details
| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752268.4 | XP_002752314.1 | 2 - 3 | 924 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPSB2| Gene Symbol: SPSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752268.4 | XP_002752314.1 | 2 - 3 | 924 | 64 | |
Set Membership: |
|