Assay Details
| Gene Symbol: LOC100395293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984156.1 | XP_008982404.1 | 4 - 5 | 2203 | 79 | |
| XM_008984157.1 | XP_008982405.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984158.2 | XP_008982406.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984159.1 | XP_008982407.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984160.1 | XP_008982408.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984162.1 | XP_008982410.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984163.1 | XP_008982411.1 | 4 - 5 | 2203 | 79 | ||
| XM_008984165.2 | XP_008982413.1 | 4 - 5 | 1703 | 79 | ||
| XM_008984166.1 | XP_008982414.1 | 8 - 9 | 1284 | 79 | ||
| XM_008984167.2 | XP_008982415.1 | 8 - 9 | 1279 | 79 | ||
| XM_008984168.2 | XP_008982416.2 | 11 - 12 | 1395 | 79 | ||
| XM_008984169.1 | XP_008982417.1 | 4 - 5 | 2203 | 79 | ||
More Information
Set Membership: |
|

LOC100395293
Genomic Map