Assay Details
| Gene Symbol: LOC100395293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984168.2 | XP_008982416.2 | 2 - 3 | 291 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100395293| Gene Symbol: LOC100395293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984168.2 | XP_008982416.2 | 2 - 3 | 291 | 63 | |
Set Membership: |
|