Assay Details
| Gene Symbol: RAD51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006223.2 | XP_009004471.1 | 2 - 3 | 319 | 92 | |
| XM_009006225.2 | XP_009004473.1 | 2 - 3 | 319 | 92 | ||
| XM_009006226.2 | XP_009004474.1 | 2 - 3 | 319 | 92 | ||
| XM_009006227.2 | XP_009004475.1 | 2 - 3 | 165 | 92 | ||
| XM_009006229.2 | XP_009004477.1 | 2 - 3 | 319 | 92 | ||
| XM_009006230.2 | XP_009004478.1 | 2 - 3 | 165 | 92 | ||
| XM_017977168.1 | XP_017832657.1 | 2 - 3 | 165 | 92 | ||
| XM_017977169.1 | XP_017832658.1 | 2 - 3 | 319 | 92 | ||
More Information
Set Membership: |
|

RAD51B
Genomic Map