Assay Details
| Gene Symbol: TNFRSF11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979926.2 | XP_008978174.1 | 5 - 6 | 588 | 67 | |
| XM_017963525.1 | XP_017819014.1 | 6 - 7 | 1251 | 67 | ||
| XM_017963526.1 | XP_017819015.1 | 5 - 6 | 1933 | 67 | ||
More Information
Set Membership: |
|

TNFRSF11A
Genomic Map