Assay Details
| Gene Symbol: C22H19orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967813.1 | XP_017823302.1 | 5 | 1755 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22H19orf48| Gene Symbol: C22H19orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967813.1 | XP_017823302.1 | 5 | 1755 | 84 | |
Set Membership: |
|