Assay Details
| Gene Symbol: DDO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971315.1 | XP_017826804.1 | 3 - 4 | 377 | 65 | |
| XM_017971316.1 | XP_017826805.1 | 2 - 3 | 274 | 65 | ||
More Information
Set Membership: |
|

DDO
Genomic Map