Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000486.2 | XP_008998734.1 | 3 - 4 | 242 | 91 | |
| XM_009000488.1 | XP_008998736.1 | 2 - 3 | 273 | 91 | ||
| XM_009000489.1 | XP_008998737.1 | 2 - 3 | 327 | 91 | ||
More Information
Set Membership: |
|

SZRD1
Genomic Map