Assay Details
| Gene Symbol: PLEKHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977802.1 | XP_017833291.1 | 7 - 8 | 620 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLEKHB1| Gene Symbol: PLEKHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977802.1 | XP_017833291.1 | 7 - 8 | 620 | 75 | |
Set Membership: |
|