Assay Details
| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999806.2 | XP_008998054.1 | 2 - 3 | 577 | 65 | |
| XM_008999807.2 | XP_008998055.1 | 2 - 3 | 1087 | 65 | ||
| XM_017973870.1 | XP_017829359.1 | 3 - 4 | 765 | 65 | ||
More Information
Set Membership: |
|

MTERF4
Genomic Map