Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755588.3 | XP_002755634.1 | 3 - 4 | 952 | 73 | |
| XM_009008572.1 | XP_009006820.1 | 3 - 4 | 558 | 73 | ||
More Information
Set Membership: |
|

CDK2AP2
Genomic Map