Assay Details
| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744114.2 | XP_002744160.1 | 5 - 6 | 1017 | 77 | |
| XM_008984260.2 | XP_008982508.1 | 4 - 5 | 940 | 77 | ||
More Information
Set Membership: |
|

CCNJL
Genomic Map