Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763262.4 | XP_002763308.1 | 12 - 13 | 1839 | 89 | |
| XM_008989871.2 | XP_008988119.1 | 12 - 13 | 1839 | 89 | ||
| XM_017968668.1 | XP_017824157.1 | 11 - 12 | 1687 | 89 | ||
More Information
Set Membership: |
|

UTP14A
Genomic Map