Assay Details
| Gene Symbol: PAFAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750458.4 | XP_002750504.1 | 4 - 5 | 686 | 77 | |
| XM_017974423.1 | XP_017829912.1 | 4 - 5 | 686 | 77 | ||
| XM_017974424.1 | XP_017829913.1 | 4 - 5 | 686 | 77 | ||
| XM_017974425.1 | XP_017829914.1 | 4 - 5 | 686 | 77 | ||
More Information
Set Membership: |
|

PAFAH2
Genomic Map