Assay Details
| Gene Symbol: LOC100396946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008177.2 | XP_009006425.1 | 1 - 2 | 178 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009008177.2 transcripts.
Set Membership: |
|

LOC100396946
Genomic Map