Assay Details
| Gene Symbol: LOC100396946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755365.3 | XP_002755411.2 | 2 - 3 | 207 | 124 | |
| XM_009008175.2 | XP_009006423.1 | 3 - 4 | 237 | 124 | ||
| XM_009008177.2 | XP_009006425.1 | 3 - 4 | 331 | 124 | ||
| XM_009008180.2 | XP_009006428.1 | 3 - 4 | 555 | 124 | ||
More Information
Set Membership: |
|

LOC100396946
Genomic Map