Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762652.3 | XP_002762698.1 | 2 - 3 | 320 | 73 | |
| XM_008988996.2 | XP_008987244.1 | 7 - 8 | 1551 | 73 | ||
| XM_008988997.2 | XP_008987245.1 | 2 - 3 | 1277 | 73 | ||
| XM_017968361.1 | XP_017823850.1 | 8 - 9 | 1726 | 73 | ||
| XM_017968362.1 | XP_017823851.1 | 8 - 9 | 1636 | 73 | ||
| XM_017968363.1 | XP_017823852.1 | 8 - 9 | 1841 | 73 | ||
| XM_017968364.1 | XP_017823853.1 | 8 - 9 | 1934 | 73 | ||
| XM_017968365.1 | XP_017823854.1 | 8 - 9 | 1701 | 73 | ||
| XM_017968366.1 | XP_017823855.1 | 2 - 3 | 1024 | 73 | ||
| XM_017968367.1 | XP_017823856.1 | 8 - 9 | 1727 | 73 | ||
More Information
Set Membership: |
|

CA5B
Genomic Map