Assay Details
| Gene Symbol: RCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988397.1 | XP_008986645.1 | 3 - 4 | 891 | 60 | |
| XM_008988398.2 | XP_008986646.1 | 3 - 4 | 725 | 60 | ||
More Information
Set Membership: |
|

RCN3
Genomic Map