Assay Details
| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755944.4 | XP_002755990.4 | 9 - 10 | 865 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS35L| Gene Symbol: VPS35L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755944.4 | XP_002755990.4 | 9 - 10 | 865 | 79 | |