Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000691.2 | XP_008998939.1 | 7 - 8 | 1122 | 67 | |
| XM_017974427.1 | XP_017829916.1 | 7 - 8 | 1123 | 67 | ||
More Information
Set Membership: |
|

TRIM63
Genomic Map