Assay Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745380.3 | XP_002745426.1 | 3 - 4 | 762 | 54 | |
| XM_008992579.2 | XP_008990827.1 | 3 - 4 | 296 | 54 | ||
| XM_008992580.2 | XP_008990828.1 | 3 - 4 | 773 | 54 | ||
| XM_017970189.1 | XP_017825678.1 | 3 - 4 | 275 | 54 | ||
| XM_017970191.1 | XP_017825680.1 | 3 - 4 | 204 | 54 | ||
More Information
Set Membership: |
|

RNF175
Genomic Map