Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988595.2 | XP_008986843.2 | 1 - 2 | 353 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
