Assay Details
| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992973.2 | XP_008991221.1 | 8 - 9 | 3590 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MMRN1| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992973.2 | XP_008991221.1 | 8 - 9 | 3590 | 65 | |
Set Membership: |
|