Assay Details
| Gene Symbol: LOC100385179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744882.4 | XP_002744928.1 | 4 - 5 | 625 | 96 | |
| XM_008991960.2 | XP_008990208.1 | 4 - 5 | 625 | 96 | ||
| XM_017969874.1 | XP_017825363.1 | 4 - 5 | 536 | 96 | ||
More Information
Set Membership: |
|

LOC100385179
Genomic Map