Assay Details
| Gene Symbol: LOC100385179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744882.4 | XP_002744928.1 | 6 - 7 | 895 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100385179| Gene Symbol: LOC100385179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744882.4 | XP_002744928.1 | 6 - 7 | 895 | 106 | |
Set Membership: |
|