Assay Details
| Gene Symbol: BIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743826.3 | XP_002743872.1 | 4 - 5 | 497 | 99 | |
| XM_008979866.2 | XP_008978114.2 | 4 - 5 | 607 | 99 | ||
| XM_017964012.1 | XP_017819501.1 | 3 - 4 | 413 | 99 | ||
More Information
Set Membership: |
|

BIK
Genomic Map