Assay Details
| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748714.2 | XP_002748760.1 | 2 - 3 | 1560 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USH1G| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748714.2 | XP_002748760.1 | 2 - 3 | 1560 | 72 | |
Set Membership: |
|