Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993058.1 | XP_008991306.1 | 2 - 3 | 280 | 68 | |
| XM_017970455.1 | XP_017825944.1 | 1 - 2 | 239 | 68 | ||
More Information
Set Membership: |
|

PLAC8
Genomic Map