Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993058.1 | XP_008991306.1 | 4 - 5 | 535 | 83 | |
| XM_017970455.1 | XP_017825944.1 | 3 - 4 | 494 | 83 | ||
More Information
Set Membership: |
|

PLAC8
Genomic Map