Assay Details
| Gene Symbol: COL21A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971184.1 | XP_017826673.1 | 17 | 1866 | 95 | |
| XM_017971185.1 | XP_017826674.1 | 30 | 3693 | 95 | ||
More Information
Set Membership: |
|

COL21A1
Genomic Map