Assay Details
| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735558.3 | XP_003735606.3 | 2 - 3 | 328 | 60 | |
| XM_017967447.1 | XP_017822936.1 | 2 - 3 | 325 | 60 | ||
| XM_017967448.1 | XP_017822937.1 | 2 - 3 | 347 | 60 | ||
| XM_017967449.1 | XP_017822938.1 | 2 - 3 | 263 | 60 | ||
More Information
Set Membership: |
|

OVOL3
Genomic Map