Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761799.3 | XP_002761845.2 | 3 - 4 | 291 | 62 | |
| XM_008987311.1 | XP_008985559.1 | 3 - 4 | 439 | 62 | ||
More Information
Set Membership: |
|

ELOF1
Genomic Map