Assay Details
| Gene Symbol: SHROOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969687.1 | XP_017825176.1 | 3 - 4 | 101 | ||
| GenBank mRNA | EF215280.1 | - | 1 - 2 | 62 | 101 | |
More Information
Set Membership: |
|

SHROOM1
Genomic Map