Assay Details
| Gene Symbol: TRIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759231.4 | XP_002759277.1 | 2 - 3 | 1231 | 88 | |
| XM_017965602.1 | XP_017821091.1 | 2 - 3 | 476 | 88 | ||
More Information
Set Membership: |
|

TRIB1
Genomic Map