Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760539.4 | XP_002760585.1 | 1 - 2 | 267 | 72 | |
| XM_008985242.2 | XP_008983490.1 | 1 - 2 | 264 | 72 | ||
More Information
Set Membership: |
|

NSL1
Genomic Map