Assay Details
| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758492.4 | XP_002758538.2 | 4 - 5 | 392 | 71 | |
| XM_008981900.2 | XP_008980148.2 | 4 - 5 | 580 | 71 | ||
| XM_008981901.2 | XP_008980149.2 | 4 - 5 | 331 | 71 | ||
| XM_017964992.1 | XP_017820481.1 | 2 - 3 | 136 | 71 | ||
| XM_017964993.1 | XP_017820482.1 | 2 - 3 | 162 | 71 | ||
| XM_017964994.1 | XP_017820483.1 | 4 - 5 | 426 | 71 | ||
| XM_017964995.1 | XP_017820484.1 | 4 - 5 | 660 | 71 | ||
| XM_017964996.1 | XP_017820485.1 | 4 - 5 | 359 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002758492.4, XM_008981900.2, XM_008981901.2, XM_017964992.1, XM_017964993.1 transcripts.
Set Membership: |
|

RPP14
Genomic Map