Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761924.3 | XP_002761970.1 | 4 - 5 | 666 | 59 | |
| XM_008987552.2 | XP_008985800.1 | 4 - 5 | 437 | 59 | ||
| XM_008987553.1 | XP_008985801.1 | 4 - 5 | 673 | 59 | ||
| XM_008987555.2 | XP_008985803.1 | 4 - 5 | 434 | 59 | ||
| XM_017967832.1 | XP_017823321.1 | 4 - 5 | 437 | 59 | ||
| XM_017967833.1 | XP_017823322.1 | 4 - 5 | 783 | 59 | ||
| XM_017967834.1 | XP_017823323.1 | 4 - 5 | 622 | 59 | ||
| XM_017967835.1 | XP_017823324.1 | 4 - 5 | 453 | 59 | ||
| XM_017967836.1 | XP_017823325.1 | 4 - 5 | 434 | 59 | ||
| XM_017967837.1 | XP_017823326.1 | 4 - 5 | 777 | 59 | ||
More Information
Set Membership: |
|

RFXANK
Genomic Map