Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756664.2 | XP_002756710.1 | 2 - 3 | 517 | 97 | |
| XM_009010387.2 | XP_009008635.1 | 2 - 3 | 221 | 97 | ||
| XM_009010388.2 | XP_009008636.1 | 3 - 4 | 302 | 97 | ||
| XM_017963214.1 | XP_017818703.1 | 2 - 3 | 517 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009010388.2 transcripts.
Set Membership: |
|

PLPP4
Genomic Map