Assay Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756664.2 | XP_002756710.1 | 6 - 7 | 969 | 54 | |
| XM_009010388.2 | XP_009008636.1 | 7 - 8 | 754 | 54 | ||
| XM_017963215.1 | XP_017818704.1 | 3 - 4 | 689 | 54 | ||
More Information
Set Membership: |
|

PLPP4
Genomic Map