Assay Details
| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757912.3 | XP_002757958.1 | 1 - 2 | 263 | 64 | |
| XM_008980953.2 | XP_008979201.1 | 1 - 2 | 263 | 64 | ||
| XM_008980954.2 | XP_008979202.1 | 1 - 2 | 263 | 64 | ||
| XM_008980955.2 | XP_008979203.1 | 1 - 2 | 263 | 64 | ||
| XM_017964448.1 | XP_017819937.1 | 1 - 2 | 263 | 64 | ||
More Information
Set Membership: |
|

RBKS
Genomic Map