Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748348.4 | XP_002748394.1 | 2 - 3 | 593 | 95 | |
| XM_008997062.2 | XP_008995310.1 | 2 - 3 | 868 | 95 | ||
More Information
Set Membership: |
|

RASL10B
Genomic Map