Assay Details
| Gene Symbol: LOC103795442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004774.2 | XP_009003022.2 | 9 - 10 | 1554 | 89 | |
| XM_017976422.1 | XP_017831911.1 | 10 - 11 | 1656 | 89 | ||
More Information
Set Membership: |
|

LOC103795442
Genomic Map