Assay Details
| Gene Symbol: PDPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734386.3 | XP_003734434.1 | 12 - 13 | 1551 | 57 | |
| XM_017978624.1 | XP_017834113.1 | 12 - 13 | 1717 | 57 | ||
More Information
Set Membership: |
|

PDPK1
Genomic Map