Assay Details
| Gene Symbol: FAM206A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743180.4 | XP_002743226.1 | 2 - 3 | 675 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM206A| Gene Symbol: FAM206A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743180.4 | XP_002743226.1 | 2 - 3 | 675 | 100 | |