Assay Details
| Gene Symbol: FAM206A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743180.4 | XP_002743226.1 | 4 - 5 | 892 | 101 | |
| XM_009000957.1 | XP_008999205.1 | 4 - 5 | 521 | 101 | ||
More Information
Set Membership: |
|

FAM206A
Genomic Map