Assay Details
| Gene Symbol: TRAPPC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761256.4 | XP_002761302.1 | 4 - 5 | 453 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC2L| Gene Symbol: TRAPPC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761256.4 | XP_002761302.1 | 4 - 5 | 453 | 81 | |
Set Membership: |
|