Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969473.1 | XP_017824962.1 | 3 - 4 | 308 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017969473.1 transcripts.
Set Membership: |
|

ZFP2
Genomic Map